Learn More
Medchemexpress LLC Boc-NH-PEG8-C2-Br | 2688072-12-8 | 99.8% | 576.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138406100MG
Encompass_Preferred
Boc-NH-PEG8-C2-Br is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-51018