Learn More
Medchemexpress LLC Boc-NH-tri-(carbonylethoxymethyl)-methane | 220431-61-8 | MFCD34178797 | 98.0% | 437.44 g/mol | C18H31NO11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW19095850MG
Encompass_Preferred
Boc-NH-Tri-(carbonylethoxymethyl)-methane is a branched PEG-based linker bearing a Boc-protected primary amine and three terminal carboxylic acid groups. It is supplied as a solid suitable for small-scale synthesis and bioconjugation workflows where selective deprotection and stable amide formation are required.
  • Boc-protected amine allows selective deprotection under mild acidic conditions.
  • Three carboxylic acid termini enable conjugation to amine-containing molecules via amide bonds.
  • Branched PEG structure provides solubility and spacer functionality in bioconjugates.
  • Available in mg-scale packaging for research and development use.
  • High purity suitable for synthetic and conjugation applications.
Catalog No. 50-002-80600