Learn More
Apexbio Technology LLC Boc-ON 58632-95-4 100g
SDP
Supplier:  Apexbio Technology LLC A7004100000
Encompass_Preferred
Boc-ON is a reagent widely employed in peptide and organic synthesis for amino group protection It introduces tert-butyloxycarbonyl (Boc) protecting groups onto amines by reacting specifically with amino functionalities temporarily masking their reactivity to prevent undesired side reactions during synthetic procedures After completion of desired synthetic steps the Boc group can be removed under mild conditions restoring free amino functionalities Boc-ON is commonly utilized in peptide synthesis and the preparation of organic molecules requiring reversible amine protection
Catalog No. 50-000-97253