Learn More
Medchemexpress LLC Boc-PEG4-C2-NHS ester | 1835759-75-5 | C20H33NO10 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1411221G
Encompass_Preferred
Boc-PEG4-C2-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are molecules composed of two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other binds to the target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
Catalog No. 50-003-55626