Learn More
Medchemexpress LLC Boc-PEG4-C2-NHS ester | 1835759-75-5 | C20H33NO10 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141122500MG
Encompass_Preferred
Boc-PEG4-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. This is achieved by connecting two different ligands with a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein intended for degradation.
  • PEG-based linker
  • Synthesize PROTACs
  • Leverages intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Connects two different ligands
Catalog No. 50-003-55031