Learn More
Medchemexpress LLC Boc-Pen(Mob)-OH | 120944-75-4 | MFCD00076975 | 99.9% | 369.48 g·mol⁻¹ | C18H27NO5S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1418151G
Encompass_Preferred
Boc-Pen(Mob)-OH is a Boc-protected phenylalanine derivative provided for research use. It serves as a protected amino acid building block for peptide synthesis and medicinal chemistry applications, enabling incorporation of a 4-methoxybenzylthio-protected penicillamine residue in synthetic sequences.
  • High purity (99.9%) suitable for research applications.
  • Molecular weight 369.48 g·mol⁻¹ and formula C18H27NO5S.
  • Provided as a 1 G packaged solid for bench-scale synthesis.
  • Documentation includes COA and SDS for quality and safety verification.
  • Useful as a building block in peptide synthesis and derivatization reactions.
Catalog No. 50-002-83565