Learn More
Medchemexpress LLC Boc-Phe(4-Br)-OH | 62129-39-9 | 99.7% | 344.20 | 10 G

Supplier: Medchemexpress LLC HYW01117810G
Boc-Phe(4-Br)-OH is a phenylalanine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Phenylalanine derivative
- Influences secretion of anabolic hormones
- Supplies fuel during exercise
- Enhances mental performance during stress-related tasks
- Prevents exercise-induced muscle damage
- Recognized as beneficial ergogenic dietary substance
- Solid appearance
- Off-white to light brown color
- Purity 99.70%
- Molecular weight 344.20
- Chemical formula C14H18BrNO4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.