Learn More
Carbosynth LLC. Boc-Phe-Ser-Arg-MCA (5 mg vial), 1ea, MW: 665.74, Molecular formula: C33H43N7O8, CAS# 73554-90-2
SDP
Supplier:  Carbosynth LLC. MFS3107V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Phenylalanyl-L-Seryl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Phe-Ser-Arg-AMC, Boc-FSR-AMC, Boc-FSR-MCA, Purity: , Grade AA, Application: Substrate for Trypsin, Tryptase, 73K Protease, and Arg-Gingipain, Storage Temperature: -20C, References: S. Iwanaga, T. Morita, H. Kato, T. Harada, N. Adachi, T. Sugo, I. Maruyama, K. Takada, T. Kimura and S. Sakakibara, KININS-II: Biochemistry, Pathophysiology, and Clinical Aspects, (S. Fujii, H. Moriya and T. Suzuki, eds.), Plenum Publishing Co., 1979. pp. 147-163, M. Muramatsu, T. Itoh, M. Takei, and K. Endo, Biol. Chem. Hoppe-Seyler, 369, 617, (1988)., A. Molla, T. Yamamoto, and H. Maeda, J. Biochem., 104, 616 (1988).
Catalog No. 50-168-5974