Learn More
Medchemexpress LLC Boc-piperazine-benzoic acid | 162046-66-4 | 97.4% | 306.36 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01324910G
Encompass_Preferred
Boc-piperazine-benzoic acid is a PROTAC linker used in the synthesis of PROTACs. Compounds synthesized with this linker, such as certain androgen receptor (AR) degraders, are highly potent and orally bioavailable. These compounds effectively reduce AR protein, suppress AR-regulated genes in tumor tissues, and inhibit tumor growth without toxicity.
  • PROTAC linker
  • Used in the synthesis of PROTACs
  • Can synthesize androgen receptor (AR) degraders
  • Resulting compounds have potential for research into prostate cancer
  • For research use only
Catalog No. 50-004-54151