Learn More
Medchemexpress LLC Boc-piperazine-benzoic acid | 162046-66-4 | 97.4% | 306.36 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013249
Encompass_Preferred
Boc-piperazine-benzoic acid is a PROTAC linker used in the synthesis of PROTACs, including the PROTAC androgen receptor (AR) degrader ARD-2128. This product is for research use only.
  • Used in the synthesis of PROTACs
  • Suitable for prostate cancer research
  • Highly potent and orally bioavailable AR degrader
  • Reduces AR protein and suppresses AR-regulated genes in tumor tissues
  • Inhibits tumor growth without toxicity
Catalog No. 50-003-66758