Learn More
Medchemexpress LLC Boc-Piperazine-OH | 77279-24-4 | 99.55% | 230.30 | 100 G

Supplier: Medchemexpress LLC HY20797100G
Boc-Piperazine-OH is a PROTAC Linker and a component in SJ46420 (HY-168635), intended for research use only. This chemical is not sold to patients. It appears as a solid below 39°C and a liquid above 39°C.
- Appearance: Solid below 39°C, liquid above 39°C
- Color: Colorless to light yellow
- Shipping: Room temperature in continental US
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.