Learn More
Medchemexpress LLC Boc-Pro-prolinal | 86925-97-5 | MFCD00890691 | 98.3% | 296.36 g/mol | C15H24N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1139515MG
Encompass_Preferred
Prolyl endopeptidase inhibitor 1 (Boc-Pro-prolinal) is a small-molecule inhibitor of prolyl endopeptidase (PEP) used in biochemical and pharmacological research. It displays high affinity (Ki = 15 nM) and has reported anti-amnesic effects in preclinical studies. The compound is supplied as a solid and as DMSO stock solutions, with recommended storage at -20°C for solid material and -80°C for long-term solvent stocks.
  • High potency against prolyl endopeptidase (Ki = 15 nM).
  • High purity (98.34%).
  • Available as solid and as DMSO stock solutions.
  • Supplied in research-scale quantities suitable for in vitro and preclinical studies.
  • Storage instructions provided for solid and solvent formats.
Catalog No. 50-002-63529