Learn More
Bachem Boc-Gln-Gly-Arg-AMC · HCl
SDP
Supplier:  Bachem 4016429.0050
Encompass_Preferred
Boc-Gln-Gly-Arg-AMC · HCl, 133448-21-2, C28H40N8O8 · HCl, Mr 653.14, 50mg. Boc-QGR-AMC, fluorogenic substrate for coagulation factor XIIa and trypsin.
Catalog No. 50-194-6929