Learn More
Medchemexpress LLC Boc-Val-Ala-PAB | 1884577-99-4 | 99.9% | 393.48 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY136547500MG
Encompass_Preferred
Boc-Val-Ala-PAB is a cathepsin cleavable ADC linker utilized in the creation of antibody-drug conjugates (ADCs). ADCs consist of an antibody to which an ADC cytotoxin is attached via an ADC linker. This product is for research use only.
  • Cathepsin cleavable linker
  • Used for making antibody-drug conjugates
  • Supports the attachment of cytotoxins to antibodies
Catalog No. 50-003-51910