Learn More
Medchemexpress LLC Boc-Val-Ala-PAB-PNP | 1884578-00-0 | ≥98.0% | 558.58 | C27H34N4O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13093250MG
Encompass_Preferred
Boc-Val-Ala-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker used to connect cytotoxic payloads to antibodies during ADC synthesis. It is supplied as a protected Val-Ala-PAB derivative with a para-nitrophenyl (PNP) activated group for conjugation chemistry and is intended for research use only.
  • Cleavable Val-Ala peptide linker for cathepsin-mediated release.
  • PNP-activated ester enables efficient coupling to payloads.
  • High purity suitable for research (≥98.0%).
  • Stable powder storage at -20°C for long-term preservation.
Catalog No. 50-002-04036