Learn More
Selleck Chemical LLC Boceprevir S3733-5mg
SDP
Supplier:  Selleck Chemical LLC S37335MG
Encompass_Preferred
Boceprevir (EBP 520 SCH 503034) is an oral direct acting hepatitis C virus (HCV) protease inhibitor with Ki value of 14 nM for NS3 It is used in combination with other antiviral agents in the treatment of chronic hepatitis C genotype 1
Catalog No. 50-310-9184