Learn More
Apexbio Technology LLC Boceprevir 394730-60-0 25mg
SDP
Supplier:  Apexbio Technology LLC A326125
Encompass_Preferred
Boceprevir is an oral inhibitor targeting hepatitis C virus (HCV) NS3 protease an enzyme essential for processing viral polyproteins during HCV replication The compound binds directly and reversibly to the NS3 enzyme s active site forming a covalent adduct through its ketoamide functionality Cell-based replicon assays demonstrated that boceprevir inhibits HCV replication in cultured hepatocytes with an IC90 of approximately 350 nM In preclinical studies boceprevir exhibited favorable selectivity towards HCV NS3 relative to human neutrophil elastase Commonly boceprevir is used in research investigating HCV replication mechanisms and NS3 enzyme function as well as in antiviral combination assays involving interferon-based regimens
Catalog No. 50-000-63191