Learn More
Medchemexpress LLC Boceprevir | 394730-60-0 | MFCD22208555 | 98.5% | 519.7 g/mol | C27H45N5O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1023710MG
Encompass_Preferred
Boceprevir is a synthetic HCV NS3/4A protease inhibitor supplied for laboratory research and analytical applications. It is provided as a high-purity solid and is available in solution formats for in vitro assays.
  • High purity suitable for reference standard and analytical use.
  • Available as a solid and as solution in DMSO for convenient dosing.
  • Suitable for biochemical and virology assays targeting HCV protease activity.
  • Prepared and quality controlled with certificate of analysis for batch verification.
Catalog No. 50-002-47611