Learn More
Medchemexpress LLC Boceprevir | 394730-60-0 | MFCD22208555 | 98.5% | 519.7 g/mol | C27H45N5O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1023725MG
Encompass_Preferred
Boceprevir is a peptidomimetic HCV NS3 protease inhibitor provided as a research-grade solid for laboratory assays and analytical applications. It is commonly used in enzymatic inhibition studies and antiviral research and is available in small pack sizes suited to analytical workflows.
  • HCV NS3 protease inhibitor mechanism.
  • High purity suitable for research applications.
  • Available in small-scale solid quantities for analytical work.
  • Recommended solvent options provided for solution preparation.
  • Suitable as an analytical standard and in vitro assay reagent.
Catalog No. 50-002-45558