Learn More
Medchemexpress LLC Boceprevir | 394730-60-0 | MFCD22208555 | 98.5% | 519.7 g/mol | C27H45N5O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1023750MG
Encompass_Preferred
Boceprevir is a peptidomimetic inhibitor of the hepatitis C virus (HCV) NS3/4A serine protease supplied as a research-grade biochemical for in vitro studies. It is provided in small pack sizes and is intended for laboratory and analytical applications.
  • Peptidomimetic NS3/4A protease inhibitor for HCV research.
  • High purity (98.5%).
  • Suitable for biochemical assays and inhibitor studies.
  • Available in small pack sizes, including 50 mg.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-46134