Learn More
Medchemexpress LLC Boceprevir (EBP 520; SCH 503034) | 394730-60-0 | MFCD16038037 | 97.8% | 519.68 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY10237
Encompass_Preferred
Boceprevir (EBP 520) is a potent, highly selective, and orally bioavailable HCV NS3 protease inhibitor with a Ki of 14 nM in enzyme assays and an EC90 of 350 nM in cell-based replicon assays. It also inhibits SARS-CoV-2 3CLpro activity. This product is for research use only and not intended for sale to patients.
  • Potent, highly selective, and orally bioavailable HCV NS3 protease inhibitor
  • Ki of 14 nM in enzyme assays
  • EC90 of 350 nM in cell-based replicon assays
  • Inhibits SARS-CoV-2 3CLpro activity
  • For research use only
Catalog No. 50-003-65743