Learn More
Medchemexpress LLC Bodipy 493/503 methyl bromide | 216434-81-0 | 98.0% | 341.01 g/mol | C14H16BBrF2N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD161410MG
Encompass_Preferred
Bodipy 493/503 methyl bromide is a green-fluorescent BODIPY dye used to stain neutral lipids and lipid droplets in live and fixed cells. It offers a sharp fluorescence peak and high quantum yield, and is relatively stable to solvent polarity and pH, enabling reliable labeling for fluorescence microscopy.
  • Stains neutral lipids and lipid droplets in live and fixed cells.
  • Maximum excitation/emission at 493/503 nm.
  • High quantum yield with a sharp fluorescence peak.
  • Relatively insensitive to polarity and pH.
  • Cell permeable for intracellular labeling.
  • Supplied as a research-grade dye suitable for microscopy applications.
Catalog No. 50-002-97155