Learn More
Cayman Chemical BODIPY 508CholSTRyl LI
SDP
Supplier:  Cayman Chemical 39684500 UG
Encompass_Preferred
A fluorescently tagged cholesterol ester; ex/em = 480/508 nm, respectively; has been used to monitor cholesterol trafficking in isolated human monocyte-derived macrophages
Catalog No. 50-291-9267