Learn More
Selleck Chemical LLC BODIPY 581/591 C11-E1473-1MG
SDP
Supplier:  Selleck Chemical LLC E14731MG
Encompass_Preferred
BODIPY 581/591 C11 is a fluorescent probe for detecting lipid peroxidation and oxidative stress in living cells It shifts emission from 591 nm (red reduced form) to 510 nm (green oxidized form) upon oxidation With high sensitivity low cytotoxicity and good light stability it is widely used to study lipid peroxidation and antioxidant properties in living cells or to detect ferroptosis by reacting with hydroxyl radicals
Catalog No. 50-313-6932