Learn More
Medchemexpress LLC BODIPY-581/591 NHS ester | 150152-70-8 | 98.67% | 489.28 | 10 MG

Supplier: Medchemexpress LLC 10MGHYD1691
BODIPY-581/591 NHS ester is a bright, red fluorescent dye used for labeling amine-containing biomolecules, with excitation at 581 nm and emission at 591 nm.
- Solid, blue to black appearance.
- Soluble in DMSO at 25 mg/mL (51.10 mM); requires ultrasonic; hygroscopic DMSO may impact solubility.
- Ships at room temperature in continental US.
- Store at -20°C, sealed, away from moisture and light.
- In solvent, store at -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.