Learn More
Medchemexpress LLC BODIPY 630/650X | 380367-48-6 | 95.5% | 660.50 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11021310MG
Encompass_Preferred
BODIPY 630/650X is a red fluorescent dye designed to target amine groups. It exhibits excitation/emission maxima of 630/650 nm. This dye is suitable for covalently labeling proteins, antibodies, or other biomolecules that contain amine groups, for detection and imaging purposes.
  • Targets amine groups
  • Exhibits excitation/emission maxima of 630/650 nm
  • Can be used for covalently labeling proteins, antibodies, or other biomolecules
  • Beneficial for imaging in living cells
  • Clearly distinguishable from background cell autofluorescence emission
Catalog No. 50-003-44044