Learn More
Cayman Chemical BODIPY 630650X succInImID5mg
SDP
Supplier:  Cayman Chemical 277585 MG
Encompass_Preferred
An amine-reactive fluorescent probe; has been used, linked to adenosine A1 receptor ligands, as a fluorescent probe to quantify ligand-receptor binding; ex/em = 630/650 nm, respectively
Catalog No. 50-293-9521