Learn More
TARGETMOL CHEMICALS INC BODIPY-Cholesterol 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T410081MG
Encompass_Preferred
BODIPY-Cholesterol (TF-Chol) is a cholesterol analog with a fluorescent BODIPY motif and cellular permeability, and is a novel BODIPY cholesterol probe.BODIPY-Cholesterol can be used to study intracellular sterol uptake and interorganelle sterol flux. Purity 98.56%
Catalog No. 50-287-6412