Learn More
Medchemexpress LLC BODIPY FL alkyne 10mg | 302795-84-2 | 10 MG

Supplier: Medchemexpress LLC HY11435410MG
BODIPY FL alkyne is a green-fluorescent borondipyrromethene dye with a terminal alkyne group for copper-catalyzed click conjugation. It is bright and photostable, emits in the fluorescein (FAM) channel, and is supplied as a purified solid suitable for labeling, probe synthesis, and fluorescence imaging.
- Molecular formula C17H18BF2N3O; molecular weight 329.15.
- Excitation ~495 nm; emission ~516 nm.
- High purity (≈98.2%) by HPLC/1H NMR.
- Terminal alkyne compatible with copper(I)-catalyzed azide-alkyne cycloaddition.
- Available in small milligram quantities for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.