Learn More
Medchemexpress LLC BODIPY FL alkyne | 1MG
SDP
Supplier:  Medchemexpress LLC HY1143541MG
Encompass_Preferred
BODIPY FL alkyne (BOD FL alkyne) is a green-fluorescent dye (Ex 503 nm Em 512 nm) a bright and photostable fluorophore which emits in fluorescein (FAM) channel BODIPY FL alkyne can be conjugated with a number of azide-containing molecules by copper-catalyzed Click Chemistry
Catalog No. 50-002-27482