Learn More
Medchemexpress LLC BODIPY FL alkyne 50mg | 302795-84-2 | 50 MG

Supplier: Medchemexpress LLC HY11435450MG
BODIPY FL alkyne is an alkyne-functionalized green-fluorescent dye used for labeling and click-chemistry conjugation in biological imaging and analytical assays. It is supplied as a light-yellow solid and is formulated for dissolution in DMSO for use as a fluorescent probe.
- Green-fluorescent alkyne dye emitting in the fluorescein range.
- High purity (≈98%).
- Molecular weight about 329.15 g·mol⁻¹ and formula C17H18BF2N3O.
- Soluble in DMSO (≈50 mg/mL; ultrasonic assistance recommended).
- Light yellow solid; handle protected from light.
- Store dry and protected from light at -20°C; in solution store at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.