Learn More
Medchemexpress LLC Bodipy FL C12 | 158757-79-0 | 99.8% | 418.33 | C23H33BF2N2O2 | 10 MG

Supplier: Medchemexpress LLC HYD157010MG
BODIPY FL C12 is a green-emitting lipophilic fluorescent dye used for labeling lipids and monitoring membrane dynamics in vitro. It provides strong absorption and sharp fluorescence suitable for microscopy and in vitro assays.
- Purity 99.79%.
- Molecular formula C23H33BF2N2O2; molecular weight 418.33 g·mol⁻¹.
- Appearance orange to red solid.
- Soluble in DMSO at 50 mg/mL; ultrasonic agitation may be required.
- Optical properties: excitation max 495 nm; emission max 516 nm.
- Storage: -20°C, protect from light; in solvent store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.