Learn More
Medchemexpress LLC BODIPY FL L-Cystine | 1180490-57-6 | 99.1% | 788.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD16055MG
Encompass_Preferred
BODIPY FL L-Cystine is a thiol-reactive, green-fluorescent dye that can be used for labeling membrane proteins, proteins with hydrophobic binding sites, or hydrophobic ligands. It has excitation at 504 nm and emission at 511 nm.
  • Thiol-reactive
  • Green-fluorescent dye
  • Suitable for labeling membrane proteins
  • Suitable for labeling proteins with hydrophobic binding sites
  • Suitable for labeling hydrophobic ligands
  • Composed of two BODIPY molecules and a cysteine residue for detecting reducing environments or the degradation process of disulfide-containing molecules
  • Can be labeled to disulfide-containing polymers (e.g., PEI or pLL)
  • Fluorescence intensity increases as disulfide bonds on the polymer are cut by a reducing environment
Catalog No. 50-004-29998