Learn More
Medchemexpress LLC BODIPY-FL NHS ester | 146616-66-2 | 99.8% | 389.16 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY110210
Encompass_Preferred
BODIPY FL NHS ester is a cell membranes-penetrable amine-reactive fluorescent probe. It has high stability and is insensitive to the polarity, pH, and type of solvent, maintaining stable fluorescence properties under different environmental conditions. It can be used for the synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay.
  • Cell membranes-penetrable amine-reactive fluorescent probe
  • Maximum excitation/emission wavelength: 502/511 nm
  • High stability and insensitive to polarity, pH, and solvent type
  • Maintains stable fluorescence properties under different environmental conditions
  • Can be used for the synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay
Catalog No. 50-003-45601