Learn More
Echelon Biosciences Research Labs BODIPY FL-PI(5)P 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs C05F650UG
Encompass
BODIPY FL PI(5)P (BODIPY FL Phosphatidylinositol 5-phosphate) is a water soluble analog of PI(5)P labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
Catalog No. 50-720-074