Learn More
Cayman Chemical BODIPY FL succInImIde esTR 5mg
SDP
Supplier:  Cayman Chemical 295085 MG
Encompass_Preferred
An amine-reactive fluorescent probe; ex/em = 502/511 nm; has been used in the synthesis of protease substrates that are non-fluorescent until unquenched following proteolytic cleavage; has been used in the synthesis of a fluorescent verapamil derivative and of fluorescent inhibitors of cholesterol absorption
Catalog No. 50-298-9168