Learn More
Echelon Biosciences Research Labs BODIPY TMR-PTDINS(4)P 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs C04M6100UG
Encompass
BODIPY TMR PI(4)P (BODIPY TMR Phosphatidylinositol 4-phosphate) is a water soluble analog of PI(4)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4-phosphate (PI(4)P) is the biosynthetic precursor to PI(4,5)P2 and has an important roles in regulating sphingomyelin and glycosphingolipid metabolism and membrane trafficking at the exit of the Golgi complex. This item is non returnable
Catalog No. 50-720-063