Learn More
Medchemexpress LLC 7-(4-(2-((1,3-Dihydroxyoctadec-4-en-2-yl)amino)-2-oxoethoxy)phenyl)-5,5-difluoro-3-(thiophen-2-yl)-5 | 571186-05-5 | 99.17% | 705.70 | 250 UG
SDP
Supplier:  Medchemexpress LLC 250UGHYD1735
Encompass_Preferred
The Golgi apparatus consists of flattened vesicles stacked together, which are round with expanded and perforated edges. BODIPY TR Ceramide is a BODIPY-labeled ceramide derivative that functions as a Golgi-specific fluorescent dye. It is synthesized in the endoplasmic reticulum and subsequently transported to the Golgi apparatus through ceramide transport protein (CERT) or vesicular translocation, enabling specific labeling. This dye allows for the visualization of individual cells with an excitation/emission (Ex/Em) of 589 nm/616 nm.
  • Golgi-specific fluorescent dye
  • Can visualize individual cells
  • Excitation/emission (Ex/Em) = 589 nm/616 nm
Catalog No. 50-004-30477