Learn More
Hach Company Boiling Chips, Silicon Carbide
Supplier:  Hach Company 2055734
Encompass
Carbon boiling chips are chemically inert to concentrated sulfuric acid, strong alkalies, and other chemicals. Carbon chips can be used for atmospheric or vacuum operations. Silicon carbide (carborundum) chips are random-sized pieces up to 2 mm long.
Catalog No. 50-199-5190