Learn More
Medchemexpress LLC Boldine | 476-70-0 | 5 MG

Supplier: Medchemexpress LLC HYN69735MG
Boldine is an apomorphine isoquinoline alkaloid extracted from the root of the pheasant pepper (Litsea cubeba). It is an orally effective antioxidant, anti-inflammatory, and antitumor agent that can inhibit osteoclast formation. Boldine induces apoptosis of human bladder cancer cells by regulating ERK, AKT, and GSK-3β, and it ameliorates bone destruction by down-regulating the OPG/RANKL/RANK signaling pathway. It can be used in rheumatoid arthritis research.
- Purity: 99.0%
- Appearance: Solid
- Color: Off-white to light yellow
- Molecular weight: 327.37
- Formula: C19H21NO4
- Structure classification: Alkaloids, isoquinoline alkaloids, phenols, polyphenols
- Initial source: Plants, Lauraceae, Litsea cubeba (Lour.) Pers.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.