Learn More
Grainger Boltless Shelving 36x84x48in AdOn
Supplier:  Grainger 14L151
Encompass_Preferred
The Grainger 14L151 is a 14-inch diameter, 3-blade, aluminum propeller for small aircraft. It has a pitch of 13 inches and is designed for engines up to 65 horsepower. The propeller is constructed from heat treated aluminum alloy for strength and durability. It uses a standard aircraft prop bolt hole pattern and includes the necessary hardware for installation. The propeller provides efficient airflow and thrust for the aircraft. With quality construction and materials, the Grainger 14L151 propeller is well-suited for use on many small piston engine aircraft.
Catalog No. 50-275-3265