Learn More
Grainger Boltless Shelving 48x84x48in Strtr
Supplier:  Grainger 14L273
Encompass_Preferred
The Edsal Boltless Shelving Unit is a 4-shelf steel shelving unit that provides versatile storage for garages, basements, workshops, and more. It features a boltless design for quick and easy assembly, with no nuts or bolts required. The unit has 4 adjustable shelves that can be moved in 1-inch increments. Each shelf holds up to 800 lbs evenly distributed. The steel construction provides strength and durability. The unit measures 36" W x 18" D x 72" H overall. Assembly hardware and instructions are included.
Catalog No. 50-274-9761