Learn More
Medchemexpress LLC Bombykol | 765-17-3 | 50 MG

Supplier: Medchemexpress LLC HYN714550MG
Bombykol is an insect sex pheromone and a sex attractant produced by female *Bombyx mandarina*. It is for research use only. When male *Bombyx mandarina* are captured in the wild using traps baited with Bombykol, they are mainly attracted in the morning, suggesting it attracts newly emerged or previously unmated males.
- Insect sex pheromone and attractant
- Produced by female *Bombyx mandarina*
- For research use only
- High purity: 99.54%
- Molecular weight: 238.41
- Chemical formula: C16H30O
- Colorless to light yellow liquid appearance
- Initial source: Silkworm moth (*Bombyx mori*)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.