Learn More
Medchemexpress LLC Bomedemstat (hydrochloride) | 99.9% | 556.07 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY109169B
Encompass_Preferred
Bomedemstat (IMG-7289) hydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It can increase H3K4 and H3K9 methylation, which then alters gene expression. Bomedemstat hydrochloride demonstrates anti-cancer activities, inhibits cancer cell proliferation, and induces apoptosis.
  • Orally active
  • Irreversible lysine-specific demethylase 1 (LSD1) inhibitor
  • Increases H3K4 and H3K9 methylation
  • Alters gene expression
  • Shows anti-cancer activities
  • Inhibits cancer cell proliferation
  • Induces apoptosis
Catalog No. 50-003-47653