Learn More
Leica Biosystems Leica BOND Covertile Cleaning Rack (S21.4588.B)
Supplier:  Leica Biosystems S21.4588.B
Encompass
The BOND Covertile cleaning rack is an accessory designed to organize and support Covertile components during cleaning and maintenance procedures for BOND instruments. It provides a purpose-built platform to streamline cleaning workflows and safe handling of parts.
  • Provides secure support for components during cleaning.
  • Organizes parts to improve workflow efficiency.
  • Durable design for routine maintenance use.
  • Easy to load and remove components for processing.
Catalog No. NC2231295