Learn More
Chemglass Life Sciences Bonded Closure, Black, 8-425, PTFE/Silicone
SDP
Supplier:  Chemglass Life Sciences CV38400008
Encompass_Preferred
Bonded Closure, Black, 8-425, PTFE/Silicone. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit design.
Catalog No. 50-231-099