Learn More
StemExpress Bone Marrow Mononuclear Cells, Frozen, 100 million/vial
SDP
Supplier:  StemExpress BMMNC100C
Encompass
Bone marrow MNCs are separated from whole bone marrow by red blood cell sedimentation followed by immunomagnetic depletion of non-MNCs. The highly pure population of MNCs is ideal for further isolations of specific cell types within the mononuclear fraction.
Catalog No. 50-212-6220