Learn More
ATCC Bone Marrow-Derived Mesenchymal Stem Cells; Normal Human
Supplier:  ATCC PCS500012
Encompass
Bone marrow-derived mesenchymal stem cells are multipotent cells isolated from the bone marrow of a donor. The cells can be used in adult stem cell differentiation, regenerative medicine, cell therapy, tissue engineering, and generation of iPS cell lines.
Catalog No. 50-238-3585
 Restricted Item