Learn More
Cayman Chemical BongkrekIc AcId 100ug
SDP
Supplier:  Cayman Chemical 38230100 UG
Encompass_Preferred
A polyketide and bacterial toxin; an inhibitor of AAT; prevents ADP import and inhibits mitochondrial respiration (Ki = 20 nM in isolated rat liver mitochondria); induces cell death in chronically estradiol-deprived MCF-7 cells (EC50 = 2.6 µM); is toxic to mice (LD50s = 0.68-6.84 mg/kg, p.o)
Catalog No. 50-291-6410