Learn More
Apexbio Technology LLC BOP reagent 56602-33-6 25g
SDP
Supplier:  Apexbio Technology LLC A701525000
Encompass_Preferred
BOP reagent is a peptide-coupling reagent designed to facilitate amide bond formation between amino acids BOP reagent acts by activating carboxyl groups on amino acid derivatives or peptides promoting coupling reactions under mild conditions The mechanism involves the generation of benzotriazole group-bearing intermediates that enable efficient coupling with amino-functionalized reactants Based on these chemical properties BOP reagent is utilized in peptide synthesis and in the preparation of phenyl ester derivatives of amino acids for research in biochemical and pharmaceutical fields
Catalog No. 50-224-0521